missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium carbonate decahydrate, 99+%, extra pure, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC271071000

 View more versions of this product

Catalog No. AC271071000



Description

Description

Specifications

Specifications

Sodium carbonate decahydrate
1.4600g/mL
1.46
White
100g
99+%
Na2CO3·10H2O
15,8731
Solubility in water: 210g/L (20°C). Other solubilities: soluble in glycerol, insoluble in alcohol
C(=O)([O-])[O-].O.O.O.O.O.O.O.O.O.O.[Na+].[Na+]
286.14
286.14
99+%
10ppm max
Plastic bottle
30ppm max
34.0°C
6132-02-1
CNa2O3·10H2O
07,322
acmc-1b9jt, carbonic acid disodium salt, decahydrate, disodium carbonate decahydrate, disodium decahydrate carbonate, ksc353a3t, natrii carbonas decahydricus, natrium-carbonat-10-wasser, natrium-carbonat-dekahydrat, sodium carbonate decahydrate, unii-ls505bg22i
XYQRXRFVKUPBQN-UHFFFAOYSA-L
disodium;carbonate;decahydrate
151402
Crystals
Extra Pure

Chemical Identifiers

6132-02-1
286.14
acmc-1b9jt, carbonic acid disodium salt, decahydrate, disodium carbonate decahydrate, disodium decahydrate carbonate, ksc353a3t, natrii carbonas decahydricus, natrium-carbonat-10-wasser, natrium-carbonat-dekahydrat, sodium carbonate decahydrate, unii-ls505bg22i
disodium;carbonate;decahydrate
CNa2O3·10H2O
XYQRXRFVKUPBQN-UHFFFAOYSA-L
151402
C(=O)([O-])[O-].O.O.O.O.O.O.O.O.O.O.[Na+].[Na+]
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Warning

SDS Documents
Product Certifications