Please login to your online account to display your discounted pricing

Sodium carbonate decahydrate, 99+%, for analysis, ACROS Organics™


Quantity 2.5kg
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  213500025

 View more versions of this product

Catalog No. AC213500025

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥99.0%
Formula Weight 286.14
Purity Grade For analysis
Physical Form Small Crystals
Quantity 2.5kg
Color White
Density 1.46g/mL
Fieser 07,322
Merck Index 15, 8731
CAS 1545739
Packaging Plastic bottle
Potassium (K) 100ppm max.
Calcium (Ca) 20ppm max.
Total Nitrogen 0.0005% max.
Assay Percent Range 99+%
Silicate and Phosphate 0.002% max.
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Chemical Name or Material Sodium carbonate decahydrate
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Linear Formula Na2CO3·10H2O
Name Note p.a.
MDL Number MFCD00149178
Molecular Formula CNa2O3·10H2O
Iron (Fe) 2ppm max.
Heavy Metals (as Pb) 10ppm max.
Magnesium (Mg) 2ppm max.
Chlorine (Cl) 5ppm max.
Aluminum (Al) 5ppm max.

Specifications

Chemical Name or Material Sodium carbonate decahydrate
Name Note p.a.
CAS 1545739
Purity Grade For analysis
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula CNa2O3·10H2O
Linear Formula Na2CO3·10H2O
Formula Weight 286.14
MDL Number MFCD00149178
Accessories

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.

SDS & Safety Documents