Please login to your online account to display your discounted pricing

Sodium Carbonate, Certified, 1.00M ±0.01M (2.0N), LabChem™

$99.91 - $99.91

Chemical Identifiers

Chemical Name or Material Sodium Carbonate
CAS 7732-18-5
CAS Max % 89.4
CAS 497-19-8
CAS Max % 10.6
Purity Grade Certified Grade
Linear Formula Na2CO3
Formula Weight 105.99
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC233902

 
lab chem
LC233902
1L Each for $99.91
Description & Specifications

Specifications

Purity Grade Certified Grade
Formula Weight 105.99
Chemical Name or Material Sodium Carbonate
Identification Passes Test
Traceability to NIST Traceable to NIST
Packaging Poly Bottle
Physical Form Liquid
Solubility Soluble in water
Specific Gravity 1g/mL
Storage Information Room Temperature
Linear Formula Na2CO3
CAS 7732-18-5
Decomposition Information Fume; Carbon monoxide; Carbon dioxide
Color Clear, colorless
Quantity 1L
Health Hazard 1 Warning
CAS Max % 89.4
Health Hazard 2 GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
CAS 497-19-8
Health Hazard 3 GHS P Statement
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
CAS Max % 10.6
Concentration 1.00M ±0.01M (2.0N)
Density 1g/mL
Safety and Handling

Warning

GHS P Statement
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.