Please login to your online account to display your discounted pricing

Sodium Carbonate, Certified, 0.530% (w/v) ±0.005%, LabChem™

$49.51 - $49.51

Chemical Identifiers

Chemical Name or Material Sodium Carbonate
CAS 7732-18-5
CAS Max % 99.47
CAS 497-19-8
CAS Max % 0.53
Purity Grade Certified Grade
Linear Formula Na2CO3
Formula Weight 105.99
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC229752

 
lab chem
LC229752
1L Each for $49.51
Description & Specifications

Specifications

Purity Grade Certified Grade
Assay 0.530% ± 0.005% (w/v)
Formula Weight 105.99
Chemical Name or Material Sodium Carbonate
Identification Passes Test
Packaging Poly Bottle
Physical Form Liquid
Solubility Soluble in water
Specific Gravity 1g/mL
Storage Information Room Temperature
Linear Formula Na2CO3
CAS 7732-18-5
Decomposition Information Fume; Carbon monoxide; Carbon dioxide
Color Clear, colorless
Quantity 1L
CAS Max % 99.47
Health Hazard 2 GHS H Statement
Solution is not hazardous.
CAS 497-19-8
Health Hazard 3 GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
CAS Max % 0.53
Density 1g/mL
Safety and Handling

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

GHS H Statement
Solution is not hazardous.