Please login to your online account to display your discounted pricing

Sodium Carbonate, Certified, 0.0250N ±0.0002N (0.0125M), LabChem™

$44.69 - $44.69

Chemical Identifiers

Chemical Name or Material Sodium Carbonate
CAS 7732-18-5
CAS Max % 99.87
CAS 497-19-8
CAS Max % 0.13
Purity Grade Certified Grade
Linear Formula Na2CO3
Formula Weight 105.99
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC230001

 
lab chem
LC230001
500mL Each for $44.69
Description & Specifications

Specifications

Purity Grade Certified Grade
Formula Weight 105.99
Chemical Name or Material Sodium Carbonate
Identification Passes Test
Packaging Poly Bottle
Physical Form Liquid
Solubility Soluble in water
Specific Gravity 1g/mL
Storage Information Room Temperature
Linear Formula Na2CO3
CAS 7732-18-5
Decomposition Information Fume; Carbon monoxide; Carbon dioxide
Color Clear, colorless
Quantity 500mL
CAS Max % 99.87
Health Hazard 2 GHS H Statement
Solution is not hazardous.
CAS 497-19-8
Health Hazard 3 GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
CAS Max % 0.13
Concentration 0.0250N ±0.0002N (0.0125M)
Density 1g/mL
Traceability to NIST Traceable to NIST
Safety and Handling

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

GHS H Statement
Solution is not hazardous.