Please login to your online account to display your discounted pricing

Sodium Carbonate, Anhydrous, ULTREX™ Ultrapure Reagent, J.T.Baker™


Quantity 100g
Packaging Wide Mouth Amber Glass Bottle

Manufacturer: avantor performance  492304

 View more versions of this product

Catalog No. 02004021

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 105.99
Chemical Name or Material Sodium Carbonate
CAS 497-19-8
Molecular Formula Na2CO3
Name Note Anhydrous
Quantity 100g
Packaging Wide Mouth Amber Glass Bottle
Purity Grade ULTREX™ Ultrapure Reagent
Specific Gravity 1L = 2.53kg

Specifications

Chemical Name or Material Sodium Carbonate
CAS 497-19-8
Name Note Anhydrous
Purity Grade ULTREX™ Ultrapure Reagent
Molecular Formula Na2CO3
Formula Weight 105.99