Please login to your online account to display your discounted pricing

Sodium Carbonate, Anhydrous, ULTREX™ Ultrapure Reagent, J.T.Baker™

$504.08 - $1,665.25

Chemical Identifiers

Chemical Name or Material Sodium Carbonate
CAS 497-19-8
Name Note Anhydrous
Purity Grade ULTREX™ Ultrapure Reagent
Molecular Formula Na2CO3
Formula Weight 105.99
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02004021

 
avantor performance
492304
Wide Mouth Amber Glass Bottle 100g Each for $1,665.25

02004022

 
avantor performance
492303
Wide Mouth Amber Glass Bottle 25g Each for $504.08
Description & Specifications

Specifications

Formula Weight 105.99
Chemical Name or Material Sodium Carbonate
CAS 497-19-8
Molecular Formula Na2CO3
Name Note Anhydrous
Quantity 100g
Packaging Wide Mouth Amber Glass Bottle
Purity Grade ULTREX™ Ultrapure Reagent
Specific Gravity 1L = 2.53kg