Please login to your online account to display your discounted pricing

Sodium Carbonate, Anhydrous, Granular, N.F. - F.C.C., J.T.Baker™

$150.40 - $682.36

Chemical Identifiers

Chemical Name or Material Sodium Carbonate
CAS 497-19-8
Name Note Anhydrous
Purity Grade FCC/NF
Molecular Formula Na2CO3
Formula Weight 105.99
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02004010

 
avantor performance
360501
Wide Mouth Amber Glass Bottle 500g Each for $150.40

02004019

 
avantor performance
360506
Poly Pail 3kg Each for $682.36
Description & Specifications

Specifications

Purity Grade FCC/NF
Formula Weight 105.99
Chemical Name or Material Sodium Carbonate
CAS 497-19-8
Name Note Anhydrous
Packaging Wide Mouth Amber Glass Bottle
Physical Form Granular
Specific Gravity 1L = 2.53kg
Molecular Formula Na2CO3
Quantity 500g