Please login to your online account to display your discounted pricing

Sodium Carbonate, Anhydrous, Granular, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$171.70 - $561.97

Chemical Identifiers

Chemical Name or Material Sodium Carbonate
CAS 497-19-8
Name Note Anhydrous
Purity Grade Baker Analyzed™ ACS Reagent
Molecular Formula Na2CO3
Formula Weight 105.99
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02004023

 
avantor performance
360401
Wide Mouth Amber Glass Bottle 500g Each for $171.70

02004024

 
avantor performance
360405
Wide Mouth Amber Glass Bottle 2.5kg Each for $561.97
Description & Specifications

Specifications

Chemical Name or Material Sodium Carbonate
Name Note Anhydrous
CAS 497-19-8
Purity Grade Baker Analyzed™ ACS Reagent
Physical Form Granular
Molecular Formula Na2CO3
Formula Weight 105.99
Packaging Wide Mouth Amber Glass Bottle
Specific Gravity 1L = 2.53kg
Quantity 500g