Please login to your online account to display your discounted pricing

Sodium Carbonate, Anhydrous, ACS Grade, ≥ 99.5%, LabChem™

$73.80 - $73.80

Chemical Identifiers

Chemical Name or Material Sodium Carbonate
CAS 497-19-8
CAS Max % 100
Name Note Anhydrous
Purity Grade ACS Grade
Linear Formula Na2CO3
Formula Weight 105.99
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC229651

 
lab chem
LC229651
500g Each for $73.80
Description & Specifications

Specifications

Assay ≥99.5%
Formula Weight 105.99
Chemical Name or Material Sodium Carbonate
EINECS Number 207-838-8
Identification Passes Test
Name Note Anhydrous
Packaging Poly Bottle
Purity Grade ACS Grade
Physical Form Powder
Solubility Soluble in water
Specific Gravity 2.53g/cm3
Storage Information Room Temperature
Linear Formula Na2CO3
CAS 497-19-8
Decomposition Information Fume; Carbon monoxide; Carbon dioxide
Color White
Quantity 500g
Health Hazard 1 Warning
CAS Max % 100
Health Hazard 2 GHS H Statement
Causes skin irritation and serious eye irritation.
Health Hazard 3 GHS P Statement
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing and wash it before reuse.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Density 2.53g/cm3
Safety and Handling

Warning

GHS P Statement
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing and wash it before reuse.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
Causes skin irritation and serious eye irritation.