missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium carbonate, anhydrous, ACS, 99.5% min. , Alfa Aesar™

Manufacturer:  Alfa Aesar 1155236

 View more versions of this product

Catalog No. AA1155236



Specifications

Specifications

Sodium carbonate, anhydrous
497-19-8
CNa2O3
White powder
CDBYLPFSWZWCQE-UHFFFAOYSA-L
disodium;carbonate
10340
105.99
ACS
Odorless
500g
For AAS analysis
99.5% min.
MFCD00003494
sodium carbonate, disodium carbonate, soda ash, carbonic acid disodium salt, calcined soda, sodium carbonate, anhydrous, carbonic acid, disodium salt, washing soda, sodiumcarbonate, solvay soda
C(=O)([O-])[O-].[Na+].[Na+]
105.988
CHEBI:29377
Granular
2.53g/cm3 at 20°C
White

Chemical Identifiers

497-19-8
105.988
CDBYLPFSWZWCQE-UHFFFAOYSA-L
10340
disodium;carbonate
CNa2O3
MFCD00003494
sodium carbonate, disodium carbonate, soda ash, carbonic acid disodium salt, calcined soda, sodium carbonate, anhydrous, carbonic acid, disodium salt, washing soda, sodiumcarbonate, solvay soda
CHEBI:29377
C(=O)([O-])[O-].[Na+].[Na+]
Safety and Handling

Safety and Handling

H319: Causes serious eye irritation.

EINECSNumber : 207-838-8

Documents