missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium carbonate, anhydrous, 99.997% (metals basis), Alfa Aesar™, Puratronic™

Manufacturer:  Thermo Fisher Scientific 1086136

 View more versions of this product

Catalog No. AA1086136



Specifications

Specifications

Sodium carbonate, anhydrous
99.997% (metals basis)
MFCD00003494
calcined soda
C(=O)([O-])[O-].[Na+].[Na+]
105.988
CHEBI:29377
Powder
White
497-19-8
CNa2O3
White powder
CDBYLPFSWZWCQE-UHFFFAOYSA-L
disodium;carbonate
10340
105.99
Odorless
500g

Chemical Identifiers

497-19-8
105.988
CDBYLPFSWZWCQE-UHFFFAOYSA-L
10340
disodium;carbonate
CNa2O3
MFCD00003494
calcined soda
CHEBI:29377
C(=O)([O-])[O-].[Na+].[Na+]
Safety and Handling

Safety and Handling

H319: Causes serious eye irritation.

EINECSNumber : 207-838-8

Documents