missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Sodium carbonate, Acculute Standard Volumetric Solution, Final Concentration 0.1N

Manufacturer:  Alfa Aesar™ 88617LQ

 View more versions of this product

Catalog No. AA88617LQ

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Sodium carbonate Acculute Standard Volumetric Solution
497-19-8
MFCD00003494
C(=O)([O-])[O-].[Na+].[Na+]
105.988
CHEBI:29377
Liquid
Final Concentration 0.1N
CNa2O3
CDBYLPFSWZWCQE-UHFFFAOYSA-L
disodium;carbonate
10340
105.99
1 unit

Chemical Identifiers

497-19-8
105.988
CDBYLPFSWZWCQE-UHFFFAOYSA-L
CHEBI:29377
C(=O)([O-])[O-].[Na+].[Na+]
CNa2O3
MFCD00003494
10340
disodium;carbonate
Safety and Handling

Safety and Handling

GHS H Statement
H319
Causes serious eye irritation.

GHS P Statement
P280-P264-P305+P351+P338-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.

Warning

TSCA : TSCA

SDS Documents