missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium carbonate, 99+%, for analysis, granular, ACROS Organics™

$109.11 - $109.11

Chemical Identifiers

CAS 497-19-8
Physical Form Fine Granular Powder
Assay Percent Range 99+%
Molecular Formula CNa2O3
Linear Formula Na2CO3
Formula Weight 105.99
MDL Number MFCD00003494
Chemical Name or Material Sodium carbonate
Assay ≥99.0%
Grade For analysis
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC206800050 SDS Thermo Fisher Scientific
AC206800050
Plastic Bucket 5kg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC206800010 SDS Thermo Fisher Scientific
206800010
Plastic Bottle 1kg Each for $109.11
AC206800025 SDS Fisher Scientific Acros
206800025
Plastic Bottle 2.5kg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Description

Biological buffer
Description & Specifications

Chemical Identifiers

497-19-8
99+%
Na2CO3
MFCD00003494
≥99.0%
Fine Granular Powder
CNa2O3
105.99
Sodium carbonate
For analysis

Specifications

Fine Granular Powder
5kg
Plastic Bucket
15, 8731
1ppm max.
300ppm max.
5ppm max.
10ppm max.
Na2CO3
0.005% max.
MFCD00003494
99+%
≥99.0%
105.99
White
07,332
20ppm max.
100ppm max.
0.002% max. (as N)
0.5% max. (285°C)
50ppm max.
20ppm max.
CNa2O3
497-19-8
Sodium carbonate
For analysis

Safety and Handling

GHS H Statement:
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning