missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sodium carbonate, 99.8%, for analysis, anhydrous, powder, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 207810050
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Adsorbent, Biological bufferSpecifications
Sodium carbonate | |
497-19-8 | |
0.5% max. (300°C) | |
Plastic Drum | |
Na2CO3 | |
5kg | |
15, 8731 | |
Solubility in water: 22 g/100mL (20°C). Other solubilities: soluble in glycerol, insoluble in alcohol | |
[Na+].[Na+].[O-]C([O-])=O | |
105.99 | |
CHEBI:29377 | |
99.8% |
Anhydrous | |
1600.0°C | |
99.8% | |
CNa2O3 | |
MFCD00003494 | |
07,332 | |
sodium carbonate, disodium carbonate, soda ash, carbonic acid disodium salt, calcined soda, sodium carbonate, anhydrous, carbonic acid, disodium salt, washing soda, sodiumcarbonate, solvay soda | |
CDBYLPFSWZWCQE-UHFFFAOYSA-L | |
disodium;carbonate | |
10340 | |
105.99 | |
Analysis |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
GHS P Statement
Wear protective gloves/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF exposed or concerned: Get med
GHS Signal Word: Warning
EINECSNumber : 207-838-8
RUO â Research Use Only