Please login to your online account to display your discounted pricing

Sodium carbonate, 99.8%, for analysis, anhydrous, powder, ACROS Organics™

$110.34 - $110.34

Chemical Identifiers

Chemical Name or Material Sodium carbonate
Name Note Anhydrous
CAS 497-19-8
Purity Grade For analysis
Physical Form Crystalline Powder or Granules
Assay ≥99.8% (on dried substance)
Assay Percent Range 99.8%
Molecular Formula CNa2O3
Linear Formula Na2CO3
Formula Weight 105.99
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC207810050

  SDS
thermo fisher scientific
AC207810050
Plastic bottle 5kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC207810010

  SDS
fisher scientific acros
207810010
Plastic bottle 1kg Each for $110.34
Description & Specifications

Specifications

Physical Form Crystalline Powder or Granules
Assay ≥99.8% (on dried substance)
Formula Weight 105.99
Quantity 5kg
Color White
Purity Grade For analysis
Fieser 07,332
Merck Index 15, 8731
Calcium (Ca) 50ppm max.
Chlorine (Cl) 10ppm max.
Linear Formula Na2CO3
Potassium (K) 50ppm max.
Trace Analysis 5ppm max.
Magnesium (Mg) 20ppm max.
Silicate 0.002% max.
Total Nitrogen Compounds 0.001% max. (as N)
Chemical Name or Material Sodium carbonate
Copper (Cu) 5ppm max.
Assay Percent Range 99.8%
Iron (Fe) 5ppm max.
Lead (Pb) 5ppm max.
Packaging Plastic bottle
Phosphate 10ppm max.
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Cadmium (Cd) 5ppm max.
Name Note Anhydrous
Molecular Formula CNa2O3
MDL Number MFCD00003494
CAS 497-19-8
Loss on Drying 0.5% max. (300°C)
Total Sulfur 0.003% max.
Aluminum (Al) 10ppm max.
Arsenic (As) 0.5ppm max.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Adsorbent, Biological buffer

Safety and Handling

Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.