Learn More
Sodium carbonate, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L130980I
Description
Sodium carbonate is commonly used as an inorganic base and a buffer component in several synthetic organic transformations in both chemical industries and research laboratories. Often used as an electrolyte, sodium carbonate acts as a very good conductor in the process of electrolysis. Apart from being used as a common chemical/biological/biotechnological laboratory reagent, it is also used as a water softener in laundering, processing wood pulp, cleaners, and making glass. It can be used for standardization of acids, and qualitative testing of cations. In dyeing with fiber-reactive dyes, sodium carbonate is used to ensure proper chemical bonding of the dye with cellulose fibers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsSodium carbonate is commonly used as an inorganic base and a buffer component in several synthetic organic transformations in both chemical industries and research laboratories. Often used as an electrolyte, sodium carbonate acts as a very good conductor in the process of electrolysis. Apart from being used as a common chemical/biological/biotechnological laboratory reagent, it is also used as a water softener in laundering, processing wood pulp, cleaners, and making glass. It can be used for standardization of acids, and qualitative testing of cations. In dyeing with fiber-reactive dyes, sodium carbonate is used to ensure proper chemical bonding of the dye with cellulose fibers.
Solubility
Soluble in water. Slightly soluble in aq. alcohol. Insoluble in carbon disulphide, acetone, and alkyl acetates.
Notes
m.p. 851° (begins to lose CO{2} at 400°)It is hygroscopic in nature, and absorbs moisture from air. On heating, it begins to lose carbon dioxide at about 400 °C.
Specifications
Sodium carbonate | |
11.5 | |
CNa2O3 | |
4154566 | |
sodium carbonate, disodium carbonate, soda ash, carbonic acid disodium salt, calcined soda, sodium carbonate, anhydrous, carbonic acid, disodium salt, washing soda, sodiumcarbonate, solvay soda | |
CDBYLPFSWZWCQE-UHFFFAOYSA-L | |
disodium;carbonate | |
10340 | |
105.99 | |
Odorless | |
Hygroscopic |
497-19-8 | |
5000 g | |
MFCD00003494 | |
14,8596 | |
Soluble in water. Slightly soluble in aq. alcohol. Insoluble in carbon disulphide,acetone,and alkyl acetates. | |
[Na+].[Na+].[O-]C([O-])=O | |
105.99 | |
CHEBI:29377 | |
98% | |
1.535 | |
2.53 |
Chemical Identifiers
497-19-8 | |
105.99 | |
CDBYLPFSWZWCQE-UHFFFAOYSA-L | |
10340 | |
[Na+].[Na+].[O-]C([O-])=O |
CNa2O3 | |
MFCD00003494 | |
sodium carbonate, disodium carbonate, soda ash, carbonic acid disodium salt, calcined soda, sodium carbonate, anhydrous, carbonic acid, disodium salt, washing soda, sodiumcarbonate, solvay soda | |
CHEBI:29377 |
Safety and Handling
GHS H Statement
H319
Causes serious eye irritation.
P234-P264b-P280i-P305+P351+P338-P390
H319
EINECSNumber : 207-838-8
RTECSNumber : VZ4050000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only