missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium Carbonate, 2.00 Normal, Ricca Chemical

Chemical Identifiers

CAS 7732-18-5,497-19-8
CAS Min % 89.0,9.4
CAS Max % 92.0,9.78
Physical Form Liquid
Chemical Name or Material Sodium Carbonate
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
719132 Ricca Chemical Company
RI719132
Poly natural 1L N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
719116 Ricca Chemical Company
RI719116
Poly natural 500mL N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

7732-18-5,497-19-8
92.0,9.78
Sodium Carbonate
89.0,9.4
Liquid

Specifications

Liquid
1L
89.0,9.4
92.0,9.78
Sodium Carbonate
Clear colorless
7732-18-5,497-19-8
2.00Normal
Poly natural