Please login to your online account to display your discounted pricing

Sodium Carbonate, 1.00 Normal, Ricca Chemical

Chemical Identifiers

Chemical Name or Material Sodium Carbonate
CAS 7732-18-5
CAS Min % 93.0
CAS Max % 97.0
CAS 497-19-8
CAS Min % 4.94
CAS Max % 5.14
Physical Form Liquid
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

719016

 
ricca chemical company
RI719016
Poly natural 500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

71902-5

 
ricca chemical company
RI71902.5
Cubitainer 10L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Color Clear colorless
Quantity 500mL
Chemical Name or Material Sodium Carbonate
CAS 7732-18-5
CAS Max % 97.0
CAS Max % 5.14
Concentration or Composition Notes 1.00Normal
CAS 497-19-8
CAS Min % 93.0
CAS Min % 4.94
Packaging Poly natural