Please login to your online account to display your discounted pricing

Sodium Carbonate, 0.100 Normal (N/10), Ricca Chemical


Quantity 20L
Packaging Cubitainer

Manufacturer: ricca chemical company  RIR718900020F

 View more versions of this product

Catalog No. R718900020F

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Color Clear colorless
Physical Form Liquid
Quantity 20L
CAS 497-19-8
Chemical Name or Material Sodium Carbonate
Solvent or Matrix Aqueous Solution
Name Note 0.100 Normal (N/10)
Assay Percent Range 0.1000 ± 0.0001 N at 20°C
CAS 7732-18-5
Odor Odorless
Packaging Cubitainer
Solubility Solubility in Water: Infinite

Specifications

Chemical Name or Material Sodium Carbonate
Name Note 0.100 Normal (N/10)
CAS 497-19-8
CAS 7732-18-5
Physical Form Liquid
Assay Percent Range 0.1000 ± 0.0001 N at 20°C