Please login to your online account to display your discounted pricing

Sodium Carbonate, 0.100 Normal (N/10), Ricca Chemical

$100.94 - $100.94

Chemical Identifiers

Chemical Name or Material Sodium Carbonate
Name Note 0.100 Normal (N/10)
CAS 497-19-8
CAS 7732-18-5
Physical Form Liquid
Assay Percent Range 0.1000 ± 0.0001 N at 20°C
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

R71890004F

 
ricca chemical company
RIR71890004F
Cubitainer 4L Each for $100.94

R718900020F

 
ricca chemical company
RIR718900020F
Cubitainer 20L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Color Clear colorless
Physical Form Liquid
Quantity 4L
CAS 497-19-8
CAS 7732-18-5
Chemical Name or Material Sodium Carbonate
Name Note 0.100 Normal (N/10)
Assay Percent Range 0.1000 ± 0.0001 N at 20°C
Odor Odorless
Packaging Cubitainer
Solubility Solubility in Water: Infinite
Solvent or Matrix Aqueous Solution