Please login to your online account to display your discounted pricing

Sodium Carbonate, 0.100 Normal (N/10), Ricca Chemical


Quantity 4L
Packaging Cubitainer

Manufacturer: ricca chemical company  RIR71890004F

 View more versions of this product

Catalog No. R71890004F

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Liquid
Color Clear colorless
Quantity 4L
CAS 497-19-8
CAS 7732-18-5
Chemical Name or Material Sodium Carbonate
Assay Percent Range 0.1000 ± 0.0001 N at 20°C
Name Note 0.100 Normal (N/10)
Packaging Cubitainer
Solubility Solubility in Water: Infinite
Odor Odorless
Solvent or Matrix Aqueous Solution

Specifications

Chemical Name or Material Sodium Carbonate
Name Note 0.100 Normal (N/10)
CAS 497-19-8
CAS 7732-18-5
Physical Form Liquid
Assay Percent Range 0.1000 ± 0.0001 N at 20°C