Please login to your online account to display your discounted pricing

Sodium Carbonate, 0.0500 Normal (N/20), 1 mL = 2.5 mg CaCO3, Ricca Chemical

Chemical Identifiers

Chemical Name or Material Sodium Carbonate
Name Note 0.0500 Normal (N/20), 1 mL = 2.5 mg CaCO3
CAS 497-19-8
CAS 7732-18-5
Assay Percent Range 0.05000 ± 0.00001 N at 20°C
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

R718500020F

 
ricca chemical company
RIR718500020F
Cubitainer 20L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

R71850004A

 
ricca chemical company
RIR71850004A
Poly natural 4L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 20L
Solvent or Matrix Aqueous Solution
Chemical Name or Material Sodium Carbonate
Name Note 0.0500 Normal (N/20), 1 mL = 2.5 mg CaCO3
CAS 497-19-8
CAS 7732-18-5
Packaging Cubitainer
Solubility Solubility in Water: Infinite
Assay Percent Range 0.05000 ± 0.00001 N at 20°C
Odor Odorless