missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sodium Carbonate, 0.0500 N (N/20), 1 mL = 2.5 mg CaCO3, Ricca Chemical
APHA/EPA for Alkalinity Testing
Supplier: Ricca Chemical Company R71850004A
Specifications
Sodium Carbonate | |
497-19-8,7732-18-5 | |
CNa2O3 | |
sodium carbonate, disodium carbonate, soda ash, carbonic acid disodium salt, calcined soda, sodium carbonate, anhydrous, carbonic acid, disodium salt, washing soda, sodiumcarbonate, solvay soda | |
[Na+].[Na+].[O-]C([O-])=O | |
105.99 | |
CHEBI:29377 | |
Natural Poly Bottle | |
4 L |
0.0500 Normal (N/20), 1 mL = 2.5 mg CaCO3 | |
0.05000 ± 0.00001 N at 20°C | |
MFCD00003494 | |
CDBYLPFSWZWCQE-UHFFFAOYSA-L | |
disodium carbonate | |
10340 | |
Odorless | |
Aqueous Solution |