Please login to your online account to display your discounted pricing

Sodium Carbonate, 0.0500 Normal (N/20), 1 mL = 2.5 mg CaCO3, Ricca Chemical


Quantity 20L
Packaging Cubitainer

Manufacturer: ricca chemical company  RIR718500020F

 View more versions of this product

Catalog No. R718500020F

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Quantity 20L
Solvent or Matrix Aqueous Solution
Chemical Name or Material Sodium Carbonate
CAS 497-19-8
Packaging Cubitainer
Solubility Solubility in Water: Infinite
CAS 7732-18-5
Name Note 0.0500 Normal (N/20), 1 mL = 2.5 mg CaCO3
Odor Odorless
Assay Percent Range 0.05000 ± 0.00001 N at 20°C

Specifications

Chemical Name or Material Sodium Carbonate
Name Note 0.0500 Normal (N/20), 1 mL = 2.5 mg CaCO3
CAS 497-19-8
CAS 7732-18-5
Assay Percent Range 0.05000 ± 0.00001 N at 20°C