Please login to your online account to display your discounted pricing

Alfa Aesar™ Sodium cacodylate, 0.1M buffer soln., pH 7.0

Chemical Identifiers

Chemical Name or Material Sodium cacodylate buffer soln.
CAS 6131-99-3
MDL Number MFCD00149079
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ60344EQE

 
alfa aesar™
J60344EQE
125mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Sodium cacodylate buffer soln.
Physical Form Liquid
MDL Number MFCD00149079
CAS 6131-99-3
pH 7.0
Color Colorless
Quantity 125mL
TSCA TSCA
Concentration 0.1M buffer soln.
Recommended Storage 2° to 8°C

Applications
Sodium cacodylate,is used as an arsinate compound for proteomics research. It is used as a buffer in electron microscopy.

Solubility
Soluble in water (~100 g/L) at 20°C, ethanol (400 mg/mL), and methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Moisture sensitive. Keep away from strong oxidizing agents, strong bases.

Safety and Handling

2° to 8°C