missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Sodium cacodylate, 0.1M buffer soln., pH 6.5

$75.72 - $138.93

Chemical Identifiers

CAS 6131-99-3
Molecular Formula C2H12AsNaO5
Molecular Weight (g/mol) 214.024
MDL Number MFCD00149079
InChI Key RLGWPHBPRCROJO-UHFFFAOYSA-M
PubChem CID 23679059
IUPAC Name sodium;dimethylarsinate;trihydrate
SMILES C[As](=O)(C)[O-].O.O.O.[Na+]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ60367AD View Documents Alfa Aesar™
J60367AD
50mL Each for $75.72
 
AAJ60367AE View Documents Alfa Aesar™
J60367AE
100mL Each for $138.93
 
Description

Description

Applications
Sodium cacodylate, is used as an arsenate compound for proteomics research. It is used as a buffer in electron microscopy.

Solubility
Soluble in water (~100 g/L) at 20°C, ethanol (400 mg/mL), and methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Moisture sensitive. Keep away from strong oxidizing agents, strong bases.
Specifications

Chemical Identifiers

6131-99-3
214.024
RLGWPHBPRCROJO-UHFFFAOYSA-M
sodium;dimethylarsinate;trihydrate
C2H12AsNaO5
MFCD00149079
23679059
C[As](=O)(C)[O-].O.O.O.[Na+]

Specifications

6131-99-3
MFCD00149079
RLGWPHBPRCROJO-UHFFFAOYSA-M
sodium;dimethylarsinate;trihydrate
23679059
6.5
Sodium cacodylate buffer soln.
C2H12AsNaO5
0.1M buffer soln.
C[As](=O)(C)[O-].O.O.O.[Na+]
214.024
Liquid
50mL
Safety and Handling

Safety and Handling

Recommended Storage : 2° to 8°C