missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium Bromide, ACS Grade, ≥ 99.0%, LabChem™

$94.51 - $94.51

Chemical Identifiers

CAS 7647-15-6
CAS Max % 100
Linear Formula NaBr
Formula Weight 102.89
Chemical Name or Material Sodium Bromide
Grade ACS Grade
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
LC229611 Lab Chem
LC229611
500g Each for $94.51
Specifications Description & Specifications

Chemical Identifiers

7647-15-6
NaBr
Sodium Bromide
100
102.89
ACS Grade

Specifications

102.89
Passes Test
Hydrogen bromide, Bromine
NaBr
Room Temperature
500g
100
Sodium Bromide
ACS Grade
Soluble in water
1,390°C
Crystalline
755°C
White
7647-15-6
Poly Bottle
≥99.0%

Safety and Handling

GHS H Statement
Product is not hazardous.

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

EINECSNumber : 231-599-9