missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Sodium bromide, 99+%, for biochemistry

CAS: 7647-15-6 | BrNa | 102.89 g/mol

$100.25 - $100.25

Chemical Identifiers

CAS 7647-15-6
Molecular Formula BrNa
Molecular Weight (g/mol) 102.89
MDL Number MFCD00003475
InChI Key JHJLBTNAGRQEKS-UHFFFAOYSA-M
Synonym sodium bromide, bromide salt of sodium, sodium bromide nabr, sedoneural, sodiumbromide, trisodium tribromide, bromnatrium, nabr, bromnatrium german, caswell no. 750a
PubChem CID 253881
ChEBI CHEBI:63004
SMILES [Na+].[Br-]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC446801000
SDS View Documents
Thermo Scientific Chemicals
446801000
100 g Plastic bottle
Each for $100.25
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7647-15-6
102.89
JHJLBTNAGRQEKS-UHFFFAOYSA-M
253881
[Na+].[Br-]
BrNa
MFCD00003475
sodium bromide, bromide salt of sodium, sodium bromide nabr, sedoneural, sodiumbromide, trisodium tribromide, bromnatrium, nabr, bromnatrium german, caswell no. 750a
CHEBI:63004

Specifications

1390.0°C
7647-15-6
99% min. (Argentometry)
NaBr
15,8729
Solubility in water: 905g/L (20°C). Other solubilities: 1160g/L water (50°C), 1210g/L water (100°C), 1g/6 mL methanol, 7g/100 mL 95% ethanol
[Na+].[Br-]
102.89
CHEBI:63004
≥99%
Plastic bottle
755.0°C
100 g
BrNa
MFCD00003475
sodium bromide, bromide salt of sodium, sodium bromide nabr, sedoneural, sodiumbromide, trisodium tribromide, bromnatrium, nabr, bromnatrium german, caswell no. 750a
JHJLBTNAGRQEKS-UHFFFAOYSA-M
sodium;bromide
253881
102.89
Biochemical
Sodium bromide
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 231-599-9

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only