missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sodium bromide, Puratronic™, 99.9955% (metals basis), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 01086022
Description
Sodium bromide is widely used for the preparation of other bromides in organic synthesis and other areas. Also used to prepare dense fluids used in oil wells.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
1390°C | |
755°C | |
White | |
100 g | |
BrNa | |
sodium bromide, bromide salt of sodium, sodium bromide nabr, sedoneural, sodiumbromide, trisodium tribromide, bromnatrium, nabr, bromnatrium german, caswell no. 750a | |
JHJLBTNAGRQEKS-UHFFFAOYSA-M | |
sodium;bromide | |
253881 | |
102.89 | |
Hygroscopic | |
3.203 |
Sodium bromide | |
7647-15-6 | |
Powder | |
99.9955% (metals basis) | |
MFCD00003475 | |
Soluble in water. Also soluble in alcohol,methanol and ethanol. | |
[Na+].[Br-] | |
102.89 | |
CHEBI:63004 | |
1.6412 | |
Powder/flake/crystalline/beads |
Chemical Identifiers
7647-15-6 | |
102.89 | |
JHJLBTNAGRQEKS-UHFFFAOYSA-M | |
253881 | |
[Na+].[Br-] |
BrNa | |
MFCD00003475 | |
sodium bromide, bromide salt of sodium, sodium bromide nabr, sedoneural, sodiumbromide, trisodium tribromide, bromnatrium, nabr, bromnatrium german, caswell no. 750a | |
CHEBI:63004 |
Safety and Handling
EINECSNumber : 231-599-9
RTECSNumber : VZ3150000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only