missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium bromide, 99.99% (metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 8760414

 View more versions of this product

Catalog No. AA8760414



Specifications

Specifications

Sodium bromide
99.99% (metals basis)
MFCD00003475
JHJLBTNAGRQEKS-UHFFFAOYSA-M
sodium;bromide
253881
102.89
Moisture sensitive
7647-15-6
BrNa
sodium bromide, bromide salt of sodium, sodium bromide nabr, sedoneural, sodiumbromide, trisodium tribromide, bromnatrium, nabr, bromnatrium german, caswell no. 750a
[Na+].[Br-]
102.894
CHEBI:63004
Crystalline
25g

Chemical Identifiers

7647-15-6
102.894
JHJLBTNAGRQEKS-UHFFFAOYSA-M
253881
sodium;bromide
BrNa
MFCD00003475
sodium bromide, bromide salt of sodium, sodium bromide nabr, sedoneural, sodiumbromide, trisodium tribromide, bromnatrium, nabr, bromnatrium german, caswell no. 750a
CHEBI:63004
[Na+].[Br-]
SDS Documents