missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium bromide, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 014037A3

 View more versions of this product

Catalog No. AA14037A3



Description

Description

Sodium bromide is used in the preparation of bromides and as a source of bromide ion.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1390°C
755°C
Granules
97%
MFCD00003475
Soluble in water,alcohol
[Na+].[Br-]
102.89
CHEBI:63004
1.6412
3.203 g/mL
Sodium bromide
7647-15-6
2 kg
BrNa
sodium bromide, bromide salt of sodium, sodium bromide nabr, sedoneural, sodiumbromide, trisodium tribromide, bromnatrium, nabr, bromnatrium german, caswell no. 750a
JHJLBTNAGRQEKS-UHFFFAOYSA-M
sodium bromide
253881
102.89
Hygroscopic

Chemical Identifiers

7647-15-6
102.89
JHJLBTNAGRQEKS-UHFFFAOYSA-M
253881
sodium bromide
BrNa
MFCD00003475
sodium bromide, bromide salt of sodium, sodium bromide nabr, sedoneural, sodiumbromide, trisodium tribromide, bromnatrium, nabr, bromnatrium german, caswell no. 750a
CHEBI:63004
[Na+].[Br-]
Safety and Handling

Safety and Handling

EINECSNumber : 231-599-9

RTECSNumber : VZ3150000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only