Please login to your online account to display your discounted pricing

Sodium Bromate (Certified), Fisher Chemical


Quantity 500g
Packaging Glass Bottle

Manufacturer: fisher scientific company  S253500

 View more versions of this product

Catalog No. S253-500

  • / Each
  • / Case of 6 Each
 

Description & Specifications

Chemical Identifiers

Melting Point 381°C
Physical Form Powder/Solid
Formula Weight 150.89
Purity Grade Certified Grade
Quantity 500g
DOT Information DOT Class 5.1, : Oxidizer
Assay 99.7 to 100.3%
pH 7
Color White
Insoluble Matter 0.01% max.
Iron (Fe) 0.002% max.
Sulfate 0.015% max.
ChemAlert Storage Symbol Yellow
Assay Percent Range 99.7 to 100.3 %
Packaging Glass Bottle
Identification Pass Test
Health Hazard 3 Emergency Overview
Cancer hazard. May cause eye, skin, and respiratory tract irritation. Use personal protective equipment. Ensure adequate ventilation. Do not breathe dust. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Get medical attention immediately if symptoms occur. Do not induce vomiting.

NFPA

Health:1
Flammability:0
Instability:0
Health Hazard 2 WARNING!
CAS 7789-38-0
Chemical Name or Material Sodium Bromate
MDL Number MFCD00003476
Heavy Metals (as Pb) 5ppm max.
Bromide P.T. (about 0.05%)
Molecular Formula BrNaO3

Specifications

Chemical Name or Material Sodium Bromate
CAS 7789-38-0
Purity Grade Certified Grade
Assay 99.7 to 100.3%
Assay Percent Range 99.7 to 100.3 %
Molecular Formula BrNaO3
Formula Weight 150.89
MDL Number MFCD00003476
Accessories
 

Emergency Overview
Cancer hazard. May cause eye, skin, and respiratory tract irritation. Use personal protective equipment. Ensure adequate ventilation. Do not breathe dust. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Get medical attention immediately if symptoms occur. Do not induce vomiting.

NFPA

Health:1
Flammability:0
Instability:0

WARNING!

SDS & Safety Documents