Please login to your online account to display your discounted pricing

Sodium Bromate (Certified), Fisher Chemical

$208.46 - $248.08

Chemical Identifiers

Chemical Name or Material Sodium Bromate
CAS 7789-38-0
Purity Grade Certified Grade
Assay 99.7 to 100.3%
Assay Percent Range 99.7 to 100.3 %
Molecular Formula BrNaO3
Formula Weight 150.89
MDL Number MFCD00003476
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

S253-250

  SDS
fisher scientific company
S253250
Glass Bottle 250g Each for $208.46
Case of 6 Each for $1,250.76

S253-500

  SDS
fisher scientific company
S253500
Glass Bottle 500g Each for $248.08
Case of 6 Each for $1,488.48
Description & Specifications

Specifications

Purity Grade Certified Grade
Physical Form Powder/Solid
Formula Weight 150.89
Melting Point 381°C
Quantity 250g
DOT Information DOT Class 5.1, : Oxidizer
Assay 99.7 to 100.3%
Viscosity [NULL]
Color White
pH 7
Identification Pass Test
Iron (Fe) 0.002% max.
Packaging Glass Bottle
Molecular Formula BrNaO3
Sulfate 0.015% max.
CAS 7789-38-0
ChemAlert Storage Symbol Yellow
Heavy Metals (as Pb) 5ppm max.
Insoluble Matter 0.01% max.
Assay Percent Range 99.7 to 100.3 %
MDL Number MFCD00003476
Bromide P.T. (about 0.05%)
Health Hazard 2 WARNING!
Health Hazard 3 Emergency Overview
Cancer hazard. May cause eye, skin, and respiratory tract irritation. Use personal protective equipment. Ensure adequate ventilation. Do not breathe dust. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Get medical attention immediately if symptoms occur. Do not induce vomiting.

NFPA

Health:1
Flammability:0
Instability:0
Chemical Name or Material Sodium Bromate
Safety and Handling
 

Emergency Overview
Cancer hazard. May cause eye, skin, and respiratory tract irritation. Use personal protective equipment. Ensure adequate ventilation. Do not breathe dust. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Get medical attention immediately if symptoms occur. Do not induce vomiting.

NFPA

Health:1
Flammability:0
Instability:0

WARNING!