Please login to your online account to display your discounted pricing

Sodium bromate, 99+%, extra pure, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  208870050

 View more versions of this product

Catalog No. AC208870050

  • / Each
 

Description & Specifications

Chemical Identifiers

Purity Grade Extra Pure
Physical Form Fine Crystalline Powder
Assay ≥99.0%
Formula Weight 150.9
Quantity 5g
Color White
EINECS Number 232-160-4
TSCA TSCA
Density 3.3300g/mL
RTECS Number EF8750000
Chemical Name or Material Sodium bromate
Name Note 99+%
Assay Percent Range 99+%
Molecular Formula BrNaO3
Linear Formula NaBrO3
Sulfate 500ppm max.
MDL Number MFCD00003476
Heavy Metals (as Pb) 50ppm max.
Iron (Fe) 50ppm max.
Packaging Glass bottle
CAS 7789-38-0
Health Hazard 1 Danger
Bromine (Br) 500ppm max.
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
May cause fire or explosion; strong oxidizer.
Health Hazard 3 GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Sodium bromate
Name Note 99+%
CAS 7789-38-0
Purity Grade Extra Pure
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula BrNaO3
Linear Formula NaBrO3
Formula Weight 150.9
MDL Number MFCD00003476
Accessories

Danger

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
May cause fire or explosion; strong oxidizer.

SDS & Safety Documents