Learn More
Sodium bromate, 99+%, extra pure, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 208875000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagentSpecifications
Sodium bromate | |
99+% | |
White | |
500 g | |
BrNaO3 | |
MFCD00003476 | |
15,8728 | |
Solubility in water: 364g/L (20°C). Other solubilities: insoluble in alcohol , ether and aceton | |
[O-]Br(=O)=O.[Na+] | |
150.9 | |
CHEBI:75229 | |
≥99% | |
Glass bottle | |
3.3300g/mL |
381.0°C | |
7789-38-0 | |
Fine Crystalline Powder | |
99+% | |
NaBrO3 | |
01,1055 | |
sodium bromate, bromic acid, sodium salt, dyetone, bromate de sodium, sodium bromate dot, neutralizer k-126, neutralizer k-140, neutralizer k-938, sodium bromate nabro3, bromate de sodium french | |
XUXNAKZDHHEHPC-UHFFFAOYSA-M | |
sodium;bromate | |
23668195 | |
150.9 | |
Extra Pure | |
3.33 |
Chemical Identifiers
7789-38-0 | |
150.9 | |
XUXNAKZDHHEHPC-UHFFFAOYSA-M | |
23668195 | |
sodium;bromate |
BrNaO3 | |
MFCD00003476 | |
sodium bromate, bromic acid, sodium salt, dyetone, bromate de sodium, sodium bromate dot, neutralizer k-126, neutralizer k-140, neutralizer k-938, sodium bromate nabro3, bromate de sodium french | |
CHEBI:75229 | |
[O-]Br(=O)=O.[Na+] |
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.
May intensify fire; oxidizer.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse caut
GHS Signal Word: Danger
EINECSNumber : 232-160-4
RTECSNumber : EF8750000
TSCA : TSCA
RUO – Research Use Only