Learn More
Sodium bromate, 99.5% (metals basis), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 01410122
Description
Sodium bromate is used as an oxidant to convert tetrahydropyranyl ethers to carbonyl compounds. It acts as an analytical reagent. It is also used for dissolving gold from its ores, boiler cleaning, oxidation of sulfur and vat dyes. Further, it is used in hair treatment formulations as a hair-permanent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Sodium bromate | |
7789-38-0 | |
Crystals | |
99.5% (metals basis) | |
MFCD00003476 | |
Soluble in water (2.5 parts in cold water,1.1 part in boiling water)Soluble in water and ammonia. Insoluble in ethanol,acetone and ether. | |
[O-]Br(=O)=O.[Na+] | |
150.891 | |
CHEBI:75229 | |
Odorless | |
White crystalline |
381°C (decomposition) | |
White | |
100 g | |
BrNaO3 | |
sodium bromate, bromic acid, sodium salt, dyetone, bromate de sodium, sodium bromate dot, neutralizer k-126, neutralizer k-140, neutralizer k-938, sodium bromate nabro3, bromate de sodium french | |
XUXNAKZDHHEHPC-UHFFFAOYSA-M | |
sodium;bromate | |
23668195 | |
150.9 | |
1.594 | |
3.34 g/cm3 |
Chemical Identifiers
7789-38-0 | |
150.891 | |
XUXNAKZDHHEHPC-UHFFFAOYSA-M | |
23668195 | |
sodium;bromate |
BrNaO3 | |
MFCD00003476 | |
sodium bromate, bromic acid, sodium salt, dyetone, bromate de sodium, sodium bromate dot, neutralizer k-126, neutralizer k-140, neutralizer k-938, sodium bromate nabro3, bromate de sodium french | |
CHEBI:75229 | |
[O-]Br(=O)=O.[Na+] |
Safety and Handling
P210-P220-P221-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
H272-H302-H315-H319-H335
DOTInformation : Hazard Class: 5.1, Packing Group: II
EINECSNumber : 232-160-4
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only