missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium bromate, 99.5% (metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 14101A3

 View more versions of this product

Catalog No. AA14101A3



Specifications

Specifications

Sodium bromate
99.5% (metals basis)
MFCD00003476
sodium bromate, bromic acid, sodium salt, dyetone, bromate de sodium, sodium bromate dot, neutralizer k-126, neutralizer k-140, neutralizer k-938, sodium bromate nabro3, bromate de sodium french
[O-]Br(=O)=O.[Na+]
150.891
CHEBI:75229
Crystals
White
7789-38-0
BrNaO3
White crystalline
XUXNAKZDHHEHPC-UHFFFAOYSA-M
sodium;bromate
23668195
150.9
Odorless
2kg

Chemical Identifiers

7789-38-0
150.891
XUXNAKZDHHEHPC-UHFFFAOYSA-M
23668195
sodium;bromate
BrNaO3
MFCD00003476
sodium bromate, bromic acid, sodium salt, dyetone, bromate de sodium, sodium bromate dot, neutralizer k-126, neutralizer k-140, neutralizer k-938, sodium bromate nabro3, bromate de sodium french
CHEBI:75229
[O-]Br(=O)=O.[Na+]
Safety and Handling

Safety and Handling

H272-H301-H315-H319: May intensify fire; oxidizer. Toxic if swallowed. Causes skin irritation. Causes serious eye irritation.

DOTInformation : Hazard Class: 5.1, Packing Group: II

EINECSNumber : 232-160-4

Documents