missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Sodium borohydride, stable aq. soln., 4.4M in 14M NaOH

Manufacturer:  Alfa Aesar™ 87659AE

 View more versions of this product

Catalog No. AA87659AE



Specifications

Specifications

Sodium borohydride
16940-66-2
NaBH4
UN3320
37.83
4.4M in 14M NaOH
Air Sensitive
100mL
Stable aq. soln.
BH4Na
MFCD00003518
Sodium tetrahydridoborate
Liquid
1.4g/mL
Colorless

Chemical Identifiers

16940-66-2
BH4Na
MFCD00003518
Sodium tetrahydridoborate
Safety and Handling

Safety and Handling

GHS H Statement
H260-H314-H302-H332
In contact with water releases flammable gases which may ignite spontaneously.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful if inhaled.

GHS P Statement
P231+P232-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Handle under inert gas.
Protect from moisture.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: SODIUM BOROHYDRIDE AND SODIUM HYDROXIDE SOLUTION

TSCA : TSCA

SDS Documents