missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium borohydride, 99%, (VenPure(TM) AF caplets), caplets, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC448495000

 View more versions of this product

Catalog No. AC448495000



Description

Description

Specifications

Specifications

Sodium borohydride
99%
NaBH4
SBH, Sodium tetrahydroborate
37.83
99%
Solubility in water: reacts. Other solubilities: soluble in ammonia - reacts with acids, soluble in amines, pyridine and dimethylether of, diethylene glycol, insol.in other ethers, hydrocarb., alkyl chlorides, reacts with oxidizing agents
500g
16940-66-2
H4BNa
15,8727
Solubility in water: reacts. Other solubilities: soluble in ammonia - reacts with acids, soluble in amines, pyridine and dimethylether of, diethylene glycol, insol.in other ethers, hydrocarb., alkyl chlorides, reacts with oxidizing agents
37.83
Glass bottle
360.0°C

Chemical Identifiers

16940-66-2
H4BNa
37.83
SBH, Sodium tetrahydroborate
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Toxic if swallowed.
May damage fertility.
May damage the unborn child.
In contact with water releases flammable gases which may ignite spontaneously.
Reacts viol

GHS P Statement
Immediately call a POISON CENTER or doctor/physician.
Keep away from any possible contact with water,because of violent reaction and possible flash fire.
IF INHALED: Remove to fresh air and keep at rest in a position comf

GHS Signal Word: Danger

EINECSNumber : 241-004-4

SDS Documents