Please login to your online account to display your discounted pricing

Sodium borohydride, 99%, VenPure™ AF caplets


Manufacturer: thermo fisher scientific  AC448490025

 View more versions of this product

Catalog No. AC448490025

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 37.83
Quantity 2.5kg
Chemical Name or Material Sodium borohydride
Assay Percent Range 99%
Molecular Formula H4BNa
Synonym SBH
CAS 16940-66-2

Specifications

Chemical Name or Material Sodium borohydride
CAS 16940-66-2
Molecular Formula H4BNa
Formula Weight 37.83
Synonym SBH