missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium borohydride, 98+%, powder, Acros Organics

Manufacturer:  Thermo Fisher Scientific 189305000

 View more versions of this product

Catalog No. AC189305000


 

Specifications Description & Specifications

Specifications

≥98.0%
37.83
White
>220°C
01,1049; 02,377; 03,262; 04,443; 05,597; 06,530; 07,329; 08,449; 09,420; 10,357; 11,477; 12,441; 13,21; 14,99; 15,290; 16,304; 17,189
16940-66-2
Sodium borohydride
98+%
Vapor Pressure: Negligible
Powder
500g
15, 8727
1.3
H4BNa
MFCD00003518
NaBH4
SBH
Glass bottle

Chemical Identifiers

16940-66-2
98+%
NaBH4
MFCD00003518
Sodium borohydride
Powder
H4BNa
37.83
SBH
≥98.0%

Safety and Handling

SDS & Safety Documents

GHS P Statement Causes severe skin burns and eye damage. Toxic if swallowed. May damage fertility. May damage the unborn child. In contact with water releases flammable gases which may ignite spontaneously. Reacts violently with water.

GHS P Statement Immediately call a POISON CENTER or doctor/physician. Keep away from any possible contact with water, because of violent reaction and possible flash fire. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF SWALLOWED: Rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Wear protective gloves/protective clothing/eye protection/face protection.

Danger

EINECSNumber : 241-004-4

RTECSNumber : ED3325000

TSCA : TSCA