Learn More
Sodium borohydride, 98+%, powder, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 189300050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagentSpecifications
Sodium borohydride | |
16940-66-2 | |
Powder | |
98% min. (Iodometry) | |
NaBH4 | |
01,1049; 02,377; 03,262; 04,443; 05,597; 06,530; 07,329; 08,449; 09,420; 10,357; 11,477; 12,441; 13,21; 14,99; 15,290; 16,304; 17,189 | |
Sodium tetrahydroborate, SBH | |
YOQDYZUWIQVZSF-UHFFFAOYSA-N | |
sodium boranuide | |
37.83 | |
Plastic Bottle | |
>220°C |
360.0°C | |
White | |
5 g | |
BH4Na | |
MFCD00003518 | |
15, 8727 | |
Solubility in water: reacts. Other solubilities: soluble in ammonia - reacts with acids, soluble in amines, pyridine and dimethylether of, diethylene glycol, insol.in other ethers, hydrocarb., alkyl chlorides, reacts with oxidizing agents | |
[BH4-].[Na+] | |
37.83 | |
≥98% | |
Negligible | |
1.3 |
Chemical Identifiers
16940-66-2 | |
37.83 | |
YOQDYZUWIQVZSF-UHFFFAOYSA-N | |
sodium boranuide |
BH4Na | |
MFCD00003518 | |
Sodium tetrahydroborate, SBH | |
[BH4-].[Na+] |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Toxic if swallowed.
May damage fertility.
May damage the unborn child.
In contact with water releases flammable gases which may ignite spontaneously.
Reacts viol
GHS P Statement
Immediately call a POISON CENTER or doctor/physician.
Keep away from any possible contact with water,because of violent reaction and possible flash fire.
IF INHALED: Remove to fresh air and keep at rest in a position comf
GHS Signal Word: Danger
EINECSNumber : 241-004-4
RTECSNumber : ED3325000
TSCA : TSCA
RUO – Research Use Only