Please login to your online account to display your discounted pricing

Sodium borohydride, 98%, granules, ca 3 mm, Acros Organics


Quantity 500g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC419475000

 View more versions of this product

Catalog No. AC419475000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5% (NaBH4)
Physical Form Granules
Formula Weight 37.83
Quantity 500g
Color White
Fieser 01,1049; 02,377; 03,262; 04,443; 05,597; 06,530; 07,329; 08,449; 09,420; 10,357; 11,477; 12,441; 13,21; 14,99; 15,290; 16,304; 17,189
Merck Index 15, 8727
UN Number 1426
Chemical Name or Material Sodium borohydride
Packaging Glass bottle
Assay Percent Range 98%
CAS 16940-66-2
Synonym SBH
Health Hazard 3 GHS P Statement Immediately call a POISON CENTER or doctor/physician. Keep away from any possible contact with water, because of violent reaction and possible flash fire. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF SWALLOWED: Rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Wear protective gloves/protective clothing/eye protection/face protection.
Molecular Formula H4BNa
Linear Formula NaBH4
MDL Number MFCD00003518
Health Hazard 1 Danger
Health Hazard 2 GHS P Statement Causes severe skin burns and eye damage. Toxic if swallowed. May damage fertility. May damage the unborn child. In contact with water releases flammable gases which may ignite spontaneously. Reacts violently with water.

Specifications

Chemical Name or Material Sodium borohydride
CAS 16940-66-2
Physical Form Granules
Assay ≥97.5% (NaBH4)
Assay Percent Range 98%
Molecular Formula H4BNa
Linear Formula NaBH4
Formula Weight 37.83
MDL Number MFCD00003518
Synonym SBH
Accessories

Danger

GHS P Statement Immediately call a POISON CENTER or doctor/physician. Keep away from any possible contact with water, because of violent reaction and possible flash fire. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF SWALLOWED: Rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Wear protective gloves/protective clothing/eye protection/face protection.

GHS P Statement Causes severe skin burns and eye damage. Toxic if swallowed. May damage fertility. May damage the unborn child. In contact with water releases flammable gases which may ignite spontaneously. Reacts violently with water.

SDS & Safety Documents