missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium Borohydride, 98%, Alfa Aesar™

Manufacturer:  Alfa Aesar 1343222

 View more versions of this product

Catalog No. AA1343222



Specifications

Specifications

Sodium Borohydride
98%
MFCD00003518
Sodium tetrahydridoborate
Caplets
220°C (428°F)
White
16940-66-2
NaBH4
White caplets
37.83
1.074g/cm3 at 20°C
Odorless
100g

Chemical Identifiers

16940-66-2
NaBH4
MFCD00003518
Sodium tetrahydridoborate
Safety and Handling

Safety and Handling

H301-H311-H331-H314-H318-H261: Toxic if swallowed. Toxic in contact with skin. Toxic if inhaled. Causes severe skin burns and eye damage. Causes serious eye damage. In contact with water releases flammable gas.

DOTInformation : DOT Class: 4.3, Packaging Group: I

EINECSNumber : 241-004-4

Documents