Please login to your online account to display your discounted pricing

Sodium borodeuteride, 98% atom D, for NMR, ACROS Organics™

$72.42 - $72.42

Chemical Identifiers

Chemical Name or Material Sodium borodeuteride
Name Note 98 atom % D
CAS 15681-89-7
Purity Grade For NMR
Assay Percent Range 98 atom% D
Molecular Formula BD4Na
Linear Formula NaBD4
Formula Weight 41.86
MDL Number MFCD00003519
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC194950010

  SDS
thermo fisher scientific
194950010
Glass bottle 1g Each for $72.42
Description & Specifications

Specifications

Purity Grade For NMR
Physical Form Powder and/or Chunks
Formula Weight 41.86
Quantity 1g
Color White to off-white to slightly gray
Fieser 03,281
EINECS Number 239-764-7
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from any possible contact with water, because of violent reaction and possible flash fire.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD00003519
Deuterium (D) 97.5% min.
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Toxic if swallowed.
Causes severe skin burns and eye damage.
In contact with water releases flammable gases which may ignite spontaneously.
Name Note 98 atom % D
CAS 15681-89-7
Chemical Name or Material Sodium borodeuteride
Linear Formula NaBD4
Assay Percent Range 98 atom% D
Molecular Formula BD4Na

For NMR

Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from any possible contact with water, because of violent reaction and possible flash fire.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Toxic if swallowed.
Causes severe skin burns and eye damage.
In contact with water releases flammable gases which may ignite spontaneously.