Please login to your online account to display your discounted pricing

Sodium Borate, Decahydrate, ACS Grade, 99.5 to 105.0%, LabChem™

$90.43 - $90.43

Chemical Identifiers

Chemical Name or Material Sodium Borate
CAS 1303-96-4
CAS Max % 100
Name Note Decahydrate
Purity Grade ACS Grade
Linear Formula Na2B4O7·10H2O
Formula Weight 381.37
Synonym Borax; Sodium Tetraborate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC229551

 
lab chem
LC229551
500g Each for $90.43
Description & Specifications

Specifications

Formula Weight 381.37
Chemical Name or Material Sodium Borate
EINECS Number 215-540-4
Identification Passes Test
Name Note Decahydrate
Packaging Poly Bottle
Purity Grade ACS Grade
Physical Form Powder
Assay 99.5 to 105.0%
Solubility Soluble in water
Specific Gravity 1.73g/cm3
Storage Information Room Temperature
Linear Formula Na2B4O7·10H2O
CAS 1303-96-4
Decomposition Information Fume; Sodium oxide; Boron
Synonym Borax; Sodium Tetraborate
Boiling Point 320°C
Color White
Melting Point 75°C
Quantity 500g
Health Hazard 1 Danger
CAS Max % 100
Health Hazard 2 GHS H Statement
May damage fertility or the unborn child.
Health Hazard 3 GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Wear eye protection, protective gloves.
If exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Density 1.73g/cm3
Safety and Handling

Danger

GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Wear eye protection, protective gloves.
If exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
May damage fertility or the unborn child.