Please login to your online account to display your discounted pricing

Sodium Borate, Certified, 40g/L, LabChem™

$63.14 - $63.14

Chemical Identifiers

Chemical Name or Material Sodium Borate
CAS 7732-18-5
CAS Max % 95.98
CAS 1303-96-4
CAS Max % 4
CAS 1310-73-2
CAS Max % 0.02
Purity Grade Certified Grade
Linear Formula Na2B4O7·10H2O
Formula Weight 381.37
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC229601

 
lab chem
LC229601
500mL Each for $63.14
Description & Specifications

Specifications

Formula Weight 381.37
pH 9.5 ± 0.1
Purity Grade Certified Grade
Specific Gravity 1g/mL
Density 1g/mL
CAS 1310-73-2
CAS Max % 4
CAS 7732-18-5
Decomposition Information Boron
Traceability to NIST Traceable to NIST
Chemical Name or Material Sodium Borate
Synonym Borax; Sodium Tetraborate
Concentration 40g/L
CAS Max % 95.98
Physical Form Liquid
Storage Information Room Temperature
Health Hazard 2 GHS H Statement
May damage fertility or the unborn child.
CAS Max % 0.02
Packaging Poly Bottle
Quantity 500mL
Color Clear, colorless
Solubility Soluble in water
Health Hazard 1 Danger
Identification Passes Test
CAS 1303-96-4
Linear Formula Na2B4O7·10H2O
Health Hazard 3 GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Wear eye protection, protective gloves.
If exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Safety and Handling

Danger

GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Wear eye protection, protective gloves.
If exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
May damage fertility or the unborn child.