Please login to your online account to display your discounted pricing

Sodium Borate Buffer Solution, LabChem™

$56.51 - $56.51

Chemical Identifiers

Chemical Name or Material Sodium Borate Buffer Solution
CAS 7732-18-5
CAS Max % 99.74
CAS 1303-96-4
CAS Max % 0.25
CAS 1310-73-2
CAS Max % 0.01
Linear Formula Na2B4O7·10H2O
Formula Weight 381.37
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC229622

 
lab chem
LC229622
1L Each for $56.51
Description & Specifications

Specifications

Formula Weight 381.37
Chemical Name or Material Sodium Borate Buffer Solution
Decomposition Information Boron
CAS Max % 0.01
Color Clear, colorless
Quantity 1L
Physical Form Liquid
Linear Formula Na2B4O7·10H2O
CAS 7732-18-5
Health Hazard 1 Danger
Identification Passes Test
CAS Max % 99.74
Health Hazard 2 GHS H Statement
May damage fertility or the unborn child.
CAS 1303-96-4
Health Hazard 3 GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Wear eye protection, protective gloves.
If exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Packaging Poly Bottle
CAS Max % 0.25
Recommended Storage Room Temperature
Solubility Soluble in water
CAS 1310-73-2
Safety and Handling

Danger

GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Wear eye protection, protective gloves.
If exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
May damage fertility or the unborn child.

Room Temperature