missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium Borate ACS AR Granular, Macron Fine Chemicals™

macron

Chemical Identifiers

CAS 1303-96-4
Physical Form Granular
Molecular Formula Na2B4O7·10H2O
Formula Weight 381.37
Chemical Name or Material Sodium Borate, 10-Hydrate
Grade ACS Grade
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
MK7457212 Avantor Performance
745706
Poly 2.5kg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

1303-96-4
Na2B4O7·10H2O
Sodium Borate, 10-Hydrate
Granular
381.37
ACS Grade

Specifications

1303-96-4
Na2B4O7·10H2O
Poly
Sodium Borate, 10-Hydrate
Granular
381.37
2.5kg
ACS Grade